This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03101210_m1
See other GOT1 GE Assays ›
Gene Symbol:
got1
Gene Name:
glutamic-oxaloacetic transaminase 1, soluble
Gene Aliases:
wu:fj63b09, zgc:55996
Chromosome Location:
Chr.13: 130154 - 137956 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03101210_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: got1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213057.2 NP_998222.1 4 - 5 665 68
XM_017358728.2 XP_017214217.1 4 - 5 661 68
GenBank mRNA BC047800.1 - 4 - 5 666 68
BC155112.1 - 4 - 5 628 68
BC164786.1 - 4 - 5 561 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
aspartate biosynthetic process
biosynthetic process
catalytic activity
L-aspartate:2-oxoglutarate aminotransferase activity
transaminase activity
transferase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0