This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03102510_m1
See other KMT5B GE Assays ›
Gene Symbol:
kmt5b
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
suv420h1, wu:fb97g06, wu:fi57g03, zgc:103527
Chromosome Location:
Chr.18: 90690 - 96322 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03102510_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: kmt5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007337.2 NP_001007338.1 5 - 6 778 73
GenBank mRNA BC085544.1 - 5 - 6 780 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
chromatin organization
muscle organ development
methylation
histone H4-K20 methylation
histone H4-K20 trimethylation
regulation of multicellular organism growth
positive regulation of isotype switching
positive regulation of double-strand break repair via nonhomologous end joining
chromatin binding
methyltransferase activity
transferase activity
histone methyltransferase activity (H4-K20 specific)
metal ion binding
S-adenosyl-L-methionine binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0