This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03104415_m1
See other GABRA1 GE Assays ›
Gene Symbol:
gabra1
Gene Name:
gamma-aminobutyric acid type A receptor subunit alpha1
Gene Aliases:
zgc:153566
Chromosome Location:
Chr.21: 42049955 - 42100776 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03104415_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gabra1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077326.1 NP_001070794.1 2 - 3 441 70
XM_009295685.3 XP_009293960.1 2 - 3 194 70
XM_009295686.3 XP_009293961.1 2 - 3 526 70
XM_009295687.3 XP_009293962.1 2 - 3 676 70
GenBank mRNA BC124697.1 - 2 - 3 441 70
BC164540.1 - 1 - 2 114 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

ion transport
chloride transport
signal transduction
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
ion transmembrane transport
regulation of membrane potential
neurological system process
synaptic transmission, GABAergic
regulation of postsynaptic membrane potential
protein heterotrimerization
chloride transmembrane transport
transmembrane signaling receptor activity
GABA-A receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
inhibitory extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
GABA-gated chloride ion channel activity
neurotransmitter receptor activity
transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline