This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03105010_m1
See other ME3 GE Assays ›
Gene Symbol:
me3
Gene Name:
malic enzyme 3, NADP(+)-dependent, mitochondrial
Gene Aliases:
im:7151680, wu:fi43b04, zgc:163117
Chromosome Location:
Chr.9: 34246384 - 34260191 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03105010_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: me3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001089356.1 NP_001082825.1 14 - 15 1818 74
GenBank mRNA BC091911.1 - 4 - 5 479 74
BC139717.1 - 14 - 15 1818 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

pyruvate metabolic process
malate metabolic process
oxidation-reduction process
malic enzyme activity
malate dehydrogenase (decarboxylating) (NAD+) activity
malate dehydrogenase (decarboxylating) (NADP+) activity
oxidoreductase activity
metal ion binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0