This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03106293_m1
See other UCP1 GE Assays ›
Gene Symbol:
ucp1
Gene Name:
uncoupling protein 1
Gene Aliases:
UCP3, UCPx, fb62c08, ucp4, wu:fb62c08, zgc:55824, zgc:77603
Chromosome Location:
Chr.1: 53374488 - 53388911 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03106293_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ucp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199523.2 NP_955817.1 6 - 7 890 54
GenBank mRNA AY398428.1 - 6 - 7 898 54
BC045464.1 - 6 - 7 898 54
BC063945.1 - 6 - 7 890 54
BC075906.1 - 6 - 7 884 54
BC164087.1 - 5 - 6 854 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
response to cold
cellular response to estrogen stimulus
cellular response to fatty acid
mitochondrial transmembrane transport
adaptive thermogenesis
oxidative phosphorylation uncoupler activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0