This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107823_m1
See other GDF11 GE Assays ›
Gene Symbol:
gdf11
Gene Name:
growth differentiation factor 11
Gene Aliases:
bmp11
Chromosome Location:
Chr.11: 257597 - 400735 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03107823_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gdf11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212975.1 NP_998140.1 2 - 3 1054 87
GenBank mRNA AF411599.3 - 2 - 3 1061 87
BC134028.1 - 2 - 3 1054 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

anterior/posterior pattern specification
positive regulation of pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
cytokine activity
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline