This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03109427_m1
See other GSR GE Assays ›
Gene Symbol:
gsr
Gene Name:
glutathione reductase
Gene Aliases:
zgc:110010
Chromosome Location:
Chr.14: 38852969 - 38873125 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03109427_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gsr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001020554.1 NP_001018390.1 10 - 11 1081 57
XM_005169592.4 XP_005169649.1 10 - 11 1200 57
XM_017359015.2 XP_017214504.1 9 - 10 1171 57
XM_021481023.1 XP_021336698.1 10 - 11 1089 57
GenBank mRNA BC095128.1 - 10 - 11 1081 57
BC165568.1 - 9 - 10 917 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
cellular response to oxidative stress
cell redox homeostasis
oxidation-reduction process
nucleotide binding
glutathione-disulfide reductase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline