This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03109528_m1
See other UCHL3 GE Assays ›
Gene Symbol:
uchl3
Gene Name:
ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase)
Gene Aliases:
im:6908825, zgc:109963
Chromosome Location:
Chr.9: 340459 - 345634 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03109528_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: uchl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024405.1 NP_001019576.1 6 - 7 588 85
GenBank mRNA BC095087.1 - 6 - 7 588 85
BC164325.1 - 6 - 7 514 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
ubiquitin-dependent protein catabolic process
Notch signaling pathway
protein deubiquitination
cranial skeletal system development
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0