Assay Details
| Gene Symbol: sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077173.1 | NP_001070641.1 | 5 - 6 | 531 | 59 | |
| XM_005158223.4 | XP_005158280.1 | 4 - 5 | 704 | 59 | ||
| GenBank mRNA | BC124210.1 | - | 5 - 6 | 531 | 59 | |
More Information
Set Membership: |
|

sybu
Genomic Map