This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03112383_m1
See other BCO1 GE Assays ›
Gene Symbol:
bco1
Gene Name:
beta-carotene oxygenase 1
Gene Aliases:
b-diox, bcdo, bcdo1, bcmo1, bcmo1a, bcox
Chromosome Location:
Chr.7: 65272470 - 65290474 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03112383_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: bco1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001328495.1 NP_001315424.1 1 - 2 193 64
GenBank mRNA AJ290390.1 - 1 - 2 162 64
BC049331.1 - 3 - 4 274 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

eye photoreceptor cell differentiation
carotene catabolic process
hindbrain development
retinal metabolic process
oxidation-reduction process
beta-carotene 15,15'-monooxygenase activity
monooxygenase activity
ferrous iron binding
carotenoid dioxygenase activity
oxidoreductase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
metal ion binding
dioxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0