This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03112718_m1
See other TP73 GE Assays ›
Gene Symbol:
tp73
Gene Name:
tumor protein p73
Gene Aliases:
TAp73, sb:eu652, zp73theta
Chromosome Location:
Chr.8: 48848200 - 48937406 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03112718_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tp73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183340.1 NP_899183.1 10 - 11 1383 72
XM_005167294.3 XP_005167351.1 11 - 12 1668 72
XM_005167295.4 XP_005167352.1 11 - 12 1653 72
XM_009304271.3 XP_009302546.1 11 - 12 1653 72
XM_009304274.3 XP_009302549.1 9 - 10 1495 72
GenBank mRNA AB199329.1 - 10 - 11 1366 72
AY233269.1 - 10 - 11 1383 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

P53-like transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
apoptotic process
cell cycle
cell cycle arrest
embryonic cranial skeleton morphogenesis
protein tetramerization
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0