This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03112734_m1
See other ALDOB GE Assays ›
Gene Symbol:
aldob
Gene Name:
aldolase b, fructose-bisphosphate
Gene Aliases:
fb25f09, wu:fb25f09
Chromosome Location:
Chr.14: 21106446 - 21115194 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03112734_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: aldob
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194367.3 NP_919348.3 8 - 9 1221 66
GenBank mRNA AF533646.1 - 8 - 9 1029 66
AY394965.1 - 8 - 9 1221 66
BC050167.1 - 8 - 9 1092 66
BC062830.1 - 8 - 9 1081 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
fructose 1,6-bisphosphate metabolic process
catalytic activity
fructose-bisphosphate aldolase activity
lyase activity
fructose-1-phosphate aldolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0