This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03114181_m1
See other PER1B GE Assays ›
Gene Symbol:
per1b
Gene Name:
period circadian clock 1b
Gene Aliases:
per4, zfper4
Chromosome Location:
Chr.7: 50822465 - 50846291 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03114181_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: per1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212439.2 NP_997604.2 12 - 13 2731 89
GenBank mRNA AY597250.1 - 12 - 13 2731 89
BC163718.1 - 11 - 12 2276 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
visual perception
response to temperature stimulus
response to light stimulus
circadian regulation of gene expression
regulation of circadian rhythm
entrainment of circadian clock by photoperiod
retina development in camera-type eye
dopaminergic neuron differentiation
positive regulation of dopaminergic neuron differentiation
transcription regulatory region sequence-specific DNA binding
transcription corepressor binding
transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline