This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03114539_m1
See other ATAT1 GE Assays ›
Gene Symbol:
atat1
Gene Name:
alpha tubulin acetyltransferase 1
Gene Aliases:
TAT, mec17, wu:fj19c03, zgc:65893, zgc:77443
Chromosome Location:
Chr.19: 27479590 - 27515653 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03114539_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: atat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001328263.1 NP_001315192.1 1 - 2 535 64
NM_001328264.1 NP_001315193.1 1 - 2 535 64
NM_213258.2 NP_998423.1 1 - 2 535 64
XM_005170018.4 XP_005170075.1 1 - 2 527 64
XM_005170020.4 XP_005170077.1 1 - 2 527 64
XM_005170021.4 XP_005170078.1 1 - 2 527 64
XM_009294208.3 XP_009292483.1 1 - 2 527 64
XM_009294209.3 XP_009292484.1 1 - 2 527 64
XM_017352219.2 XP_017207708.1 1 - 2 527 64
GenBank mRNA BC056749.1 - 1 - 2 287 64
BC065981.1 - 1 - 2 289 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neuron development
regulation of microtubule cytoskeleton organization
alpha-tubulin acetylation
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
transferase activity
transferase activity, transferring acyl groups
tubulin N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0