This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03116096_m1
See other ADGRG1 GE Assays ›
Gene Symbol:
adgrg1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
fc49b10, gpr56, wu:fc49b10
Chromosome Location:
Chr.7: 28670462 - 28696716 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03116096_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: adgrg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001020481.1 NP_001018317.1 11 - 12 1543 84
XM_005166389.4 XP_005166446.1 12 - 13 1890 84
GenBank mRNA AB193557.1 - 11 - 12 1543 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
oligodendrocyte development
peripheral nervous system myelin maintenance
regulation of apoptotic process
myelination of posterior lateral line nerve axons
regulation of oligodendrocyte progenitor proliferation
protein kinase activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0