Assay Details
| Gene Symbol: gstp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131734.3 | NP_571809.1 | 3 - 4 | 218 | 74 | |
| GenBank mRNA | AB194127.1 | - | 3 - 4 | 173 | 74 | |
| AF285098.1 | - | 3 - 4 | 173 | 74 | ||
| BC162358.1 | - | 3 - 4 | 273 | 74 | ||
| BC162376.1 | - | 3 - 4 | 273 | 74 | ||
| CK361426.1 | - | 3 - 4 | 258 | 74 | ||
More Information
Set Membership: |
|

gstp1
Genomic Map