This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03119676_m1
See other YIF1A GE Assays ›
Gene Symbol:
yif1a
Gene Name:
Yip1 interacting factor homolog A (S. cerevisiae)
Gene Aliases:
fa11c09, fk27h03, wu:fa11c09, wu:fk27h03, zgc:73136
Chromosome Location:
Chr.14: 30751403 - 30759459 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03119676_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: yif1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199931.1 NP_956225.1 6 - 7 544 63
XM_009291093.2 XP_009289368.1 7 - 8 691 63
XM_017358991.1 XP_017214480.1 6 - 7 637 63
GenBank mRNA BC046046.1 - 5 - 6 615 63
BC059499.1 - 6 - 7 544 63
BC071433.1 - 6 - 7 556 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0