This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03119710_m1
See other SLC1A2B GE Assays ›
Gene Symbol:
slc1a2b
Gene Name:
solute carrier family 1 member 2b
Gene Aliases:
eaat2, fj34b12, slc1a2, wu:fj34b12, zgc:65897
Chromosome Location:
Chr.25: 37366698 - 37407757 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03119710_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc1a2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199979.2 NP_956273.2 4 - 5 802 81
XM_009298220.3 XP_009296495.1 4 - 5 587 81
XM_009298221.3 XP_009296496.1 4 - 5 566 81
XM_009298222.3 XP_009296497.1 4 - 5 753 81
GenBank mRNA BC056751.1 - 4 - 5 727 81
BC155822.1 - 4 - 5 753 81
HM138691.1 - 3 - 4 544 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

larval locomotory behavior
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline