Assay Details
| Gene Symbol: arhgap4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200444.1 | NP_956738.1 | 19 - 20 | 2158 | 83 | |
| XM_005162155.4 | XP_005162212.1 | 19 - 20 | 2258 | 83 | ||
| GenBank mRNA | BC054931.1 | - | 19 - 20 | 2158 | 83 | |
More Information
Set Membership: |
|

arhgap4a
Genomic Map