This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03121196_m1
See other RBPMS2A GE Assays ›
Gene Symbol:
rbpms2a
Gene Name:
RNA binding protein, mRNA processing factor 2a
Gene Aliases:
zgc:92689
Chromosome Location:
Chr.25: 6122920 - 6152783 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03121196_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rbpms2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002409.1 NP_001002409.1 4 - 5 585 63
XM_005162967.4 XP_005163024.1 4 - 5 605 63
XM_009297758.3 XP_009296033.1 4 - 5 605 63
XM_021470473.1 XP_021326148.1 4 - 5 421 63
XM_021470474.1 XP_021326149.1 4 - 5 418 63
XM_021470475.1 XP_021326150.1 4 - 5 418 63
XM_021470476.1 XP_021326151.1 4 - 5 414 63
GenBank mRNA BC076171.1 - 4 - 5 585 63
BC152178.1 - 4 - 5 505 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

retinal ganglion cell axon guidance
dendrite morphogenesis
negative regulation of smooth muscle cell differentiation
regulation of retinal ganglion cell axon guidance
nucleic acid binding
RNA binding
mRNA binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline