This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03121198_m1
See other RBPMS2A GE Assays ›
Gene Symbol:
rbpms2a
Gene Name:
RNA binding protein, mRNA processing factor 2a
Gene Aliases:
zgc:92689
Chromosome Location:
Chr.25: 6122920 - 6152783 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03121198_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rbpms2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002409.1 NP_001002409.1 6 - 7 880 86
XM_021470474.1 XP_021326149.1 6 - 7 713 86
XM_021470476.1 XP_021326151.1 6 - 7 709 86
GenBank mRNA BC076171.1 - 6 - 7 880 86
BC152178.1 - 6 - 7 800 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

retinal ganglion cell axon guidance
dendrite morphogenesis
negative regulation of smooth muscle cell differentiation
regulation of retinal ganglion cell axon guidance
nucleic acid binding
RNA binding
mRNA binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0