This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03121460_m1
See other PLTP GE Assays ›
Gene Symbol:
pltp
Gene Name:
phospholipid transfer protein
Gene Aliases:
zgc:100903
Chromosome Location:
Chr.6: 55357155 - 55393321 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03121460_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pltp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003519.1 NP_001003519.1 4 - 5 422 66
XM_005168776.4 XP_005168833.1 4 - 5 469 66
GenBank mRNA BC077135.1 - 4 - 5 422 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid transport
high-density lipoprotein particle remodeling
ceramide transport
lipid binding
phosphatidylethanolamine binding
phosphatidylcholine transporter activity
diacylglycerol binding
phosphatidylcholine binding
phosphatidic acid binding
ceramide binding
intermembrane ceramide transfer activity
phosphatidylglycerol binding
phosphatidylethanolamine transporter activity
phosphatidic acid transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline