This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03123353_m1
See other SLCO1C1 GE Assays ›
Gene Symbol:
slco1c1
Gene Name:
solute carrier organic anion transporter family, member 1C1
Gene Aliases:
si:dkey-217k21.8
Chromosome Location:
Chr.4: 2674761 - 2745425 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03123353_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slco1c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044997.3 NP_001038462.2 5 - 6 670 59
XM_005164471.4 XP_005164528.1 6 - 7 1075 59
XM_009299972.3 XP_009298247.1 6 - 7 1083 59
XM_009299973.3 XP_009298248.1 6 - 7 1213 59
XM_009299975.3 XP_009298250.1 6 - 7 1086 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

ion transport
sodium-independent organic anion transport
transmembrane transport
sodium-independent organic anion transmembrane transporter activity
transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0