This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03125005_m1
See other UCP2 GE Assays ›
Gene Symbol:
ucp2
Gene Name:
uncoupling protein 2
Gene Aliases:
cb74
Chromosome Location:
Chr.10: 36662671 - 36673377 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03125005_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ucp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131176.1 NP_571251.1 3 - 4 464 60
GenBank mRNA AJ243250.1 - 3 - 4 437 60
BC056737.1 - 3 - 4 459 60
BC065607.1 - 3 - 4 464 60
BC152646.1 - 3 - 4 473 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
response to cold
mitochondrial transmembrane transport
adaptive thermogenesis
oxidative phosphorylation uncoupler activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-9bd97d8-bn4qx:80/100.66.78.49:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline