This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03125731_m1
See other GTF2B GE Assays ›
Gene Symbol:
gtf2b
Gene Name:
general transcription factor IIB
Gene Aliases:
wu:fc44c11, zgc:66258
Chromosome Location:
Chr.2: 22480115 - 22492289 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03125731_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gtf2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199697.1 NP_955991.1 5 - 6 564 83
GenBank mRNA BC055569.1 - 5 - 6 564 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcriptional start site selection at RNA polymerase II promoter
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
RNA polymerase II transcriptional preinitiation complex assembly
transcription preinitiation complex assembly
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, core RNA polymerase II recruiting
transcription factor binding
TBP-class protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline