This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03126811_m1
See other DISP1 GE Assays ›
Gene Symbol:
disp1
Gene Name:
dispatched homolog 1 (Drosophila)
Gene Aliases:
con, zgc:111866
Chromosome Location:
Chr.20: 51489195 - 51642384 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03126811_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: disp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212800.1 NP_997965.1 3 - 4 596 84
XM_017352523.2 XP_017208012.1 3 - 4 634 84
GenBank mRNA AY512779.1 - 3 - 4 596 84
BC163557.1 - 2 - 3 498 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s)

smoothened signaling pathway
multicellular organism development
skeletal muscle tissue development
blood circulation
organ morphogenesis
anterior/posterior pattern specification
adenohypophysis development
retinal ganglion cell axon guidance
muscle cell fate specification
embryonic cranial skeleton morphogenesis
inner ear development

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0