This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03128606_m1
See other CPSF6 GE Assays ›
Gene Symbol:
cpsf6
Gene Name:
cleavage and polyadenylation specific factor 6
Gene Aliases:
wu:fa22f12, zgc:85819
Chromosome Location:
Chr.4: 2043987 - 2059304 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03128606_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cpsf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017993.1 NP_001017993.1 1 - 2 70 85
XM_005164451.4 XP_005164508.1 1 - 2 132 85
XM_005164452.4 XP_005164509.1 1 - 2 132 85
XM_005164453.4 XP_005164510.1 1 - 2 132 85
GenBank mRNA AY398330.1 - 1 - 2 70 85
BC067642.1 - 1 - 2 86 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
positive regulation of RNA export from nucleus
protein heterotetramerization
mRNA alternative polyadenylation
messenger ribonucleoprotein complex assembly
nucleic acid binding
RNA binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline