This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03130207_m1
See other CD99 GE Assays ›
Gene Symbol:
cd99
Gene Name:
CD99 molecule
Gene Aliases:
im:7159661, zgc:153928
Chromosome Location:
Chr.1: 33355377 - 33380072 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03130207_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cd99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079980.1 NP_001073449.1 2 - 3 115 57
XM_005159923.4 XP_005159980.1 2 - 3 199 57
XM_005159924.4 XP_005159981.1 2 - 3 199 57
XM_005159925.4 XP_005159982.1 2 - 3 173 57
XM_009292151.3 XP_009290426.1 2 - 3 199 57
GenBank mRNA BC125914.1 - 2 - 3 115 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

homotypic cell-cell adhesion
T cell extravasation
positive regulation of neutrophil extravasation
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline