This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03131529_g1
See other ELAVL3 GE Assays ›
Gene Symbol:
elavl3
Gene Name:
ELAV like neuron-specific RNA binding protein 3
Gene Aliases:
HuC, elrc, id:ibd1248, wu:fb77b03, zHuC
Chromosome Location:
Chr.3: 46798511 - 46817871 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03131529_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: elavl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131449.1 NP_571524.1 2 - 3 253 81
XM_005164218.4 XP_005164275.1 2 - 3 626 81
XM_005164219.4 XP_005164276.1 2 - 3 626 81
XM_005164220.4 XP_005164277.1 2 - 3 626 81
XM_009299733.3 XP_009298008.1 2 - 3 626 81
XM_017353188.2 XP_017208677.1 2 - 3 626 81
XM_017353190.2 XP_017208679.1 2 - 3 626 81
XM_017353191.2 XP_017208680.1 2 - 3 626 81
GenBank mRNA BC065343.1 - 2 - 3 593 81
U62018.1 - 2 - 3 253 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of neurogenesis
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0