This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03131810_m1
See other SPNS1 GE Assays ›
Gene Symbol:
spns1
Gene Name:
spinster homolog 1 (Drosophila)
Gene Aliases:
etID64740.3, nrs, spinl, wu:fb95b12, wu:fi37e11
Chromosome Location:
Chr.3: 110201 - 130131 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03131810_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: spns1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153663.2 NP_705949.2 7 - 8 1004 62
XM_005163832.4 XP_005163889.1 7 - 8 1018 62
GenBank mRNA AF465772.1 - 7 - 8 960 62
BC048024.1 - 7 - 8 959 62
BC165625.1 - 6 - 7 804 62
CK027241.1 - 2 - 3 214 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
autophagy
transmembrane transport
cellular senescence
autophagosome maturation
transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline