This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03133428_m1
See other TAL1 GE Assays ›
Gene Symbol:
tal1
Gene Name:
T-cell acute lymphocytic leukemia 1
Gene Aliases:
SCL-alpha, SCL-beta, scl, scl/tal1, tal-1, zgc:85605
Chromosome Location:
Chr.22: 16535585 - 16545329 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03133428_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tal1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213237.1 NP_998402.1 3 - 4 686 72
GenBank mRNA AF038873.1 - 2 - 3 562 72
AF045432.2 - 3 - 4 675 72
BC068324.1 - 3 - 4 686 72
EF488003.1 - 2 - 3 263 72
EF488004.1 - 3 - 4 701 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
blood vessel development
vasculogenesis
heart morphogenesis
endocardium morphogenesis
regulation of transcription from RNA polymerase II promoter
multicellular organism development
heart development
stem cell population maintenance
hemopoiesis
myeloid cell differentiation
cell differentiation
erythrocyte differentiation
thyroid gland development
embryonic heart tube development
embryonic hemopoiesis
positive regulation of endothelial cell differentiation
lateral mesoderm development
nucleate erythrocyte development
artery morphogenesis
endocardium formation
primitive hemopoiesis
definitive hemopoiesis
hemangioblast cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-859c6f577-2spj5:80/100.66.76.192:80.
git-commit: 13ffd1f44fd3b57634547fe5abf56598aa1cdbac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.0-2026.02.07-1.0