This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03136396_m1
See other STRA6 GE Assays ›
Gene Symbol:
stra6
Gene Name:
signaling receptor and transporter of retinol STRA6
Gene Aliases:
im:7151282, wu:fc51h06, zgc:136689
Chromosome Location:
Chr.25: 22571678 - 22634675 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03136396_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: stra6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045312.1 NP_001038777.1 8 - 9 1037 72
XM_005170689.4 XP_005170746.1 9 - 10 916 72
XM_021470528.1 XP_021326203.1 9 - 10 960 72
GenBank mRNA BC117633.1 - 8 - 9 1037 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

retinol transport
chordate embryonic development
vitamin A import
calmodulin binding
retinal binding
retinol binding
retinol transporter activity
signaling receptor activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0