This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03139081_m1
See other NEIL1 GE Assays ›
Gene Symbol:
neil1
Gene Name:
nei-like DNA glycosylase 1
Gene Aliases:
zgc:56361
Chromosome Location:
Chr.25: 26827140 - 26833102 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03139081_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: neil1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200283.2 NP_956577.2 2 - 3 524 80
GenBank mRNA AY398387.1 - 1 - 2 469 80
BC049408.1 - 2 - 3 497 80
EH450678.1 - 2 - 3 525 80
EH538356.1 - 2 - 3 548 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

base-excision repair
nucleic acid binding
damaged DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
nuclease activity
endonuclease activity
zinc ion binding
hydrolase activity
hydrolase activity, hydrolyzing N-glycosyl compounds
DNA N-glycosylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline