This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03139193_m1
See other DTX3 GE Assays ›
Gene Symbol:
dtx3
Gene Name:
deltex E3 ubiquitin ligase 3
Gene Aliases:
deltex3, fi25g12, wu:fi25g12, zgc:66440
Chromosome Location:
Chr.23: 27040128 - 27050114 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03139193_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dtx3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200528.2 NP_956822.2 3 - 4 1476 112
XM_005162242.4 XP_005162299.1 4 - 5 1501 112
XM_005162244.4 XP_005162301.1 3 - 4 1142 112
XM_009296836.3 XP_009295111.1 3 - 4 1470 112
XM_009296837.3 XP_009295112.1 4 - 5 1090 112
XM_009296838.3 XP_009295113.1 5 - 6 1149 112
XM_017353528.1 XP_017209017.1 3 - 4 1388 112
GenBank mRNA BC055658.1 - 3 - 4 1078 112
BC098516.1 - 3 - 4 1476 112
DQ497601.1 - 2 - 3 998 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

Notch signaling pathway
protein ubiquitination
metal ion binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0