This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03139750_m1
See other EIF2S3 GE Assays ›
Gene Symbol:
eif2s3
Gene Name:
eukaryotic translation initiation factor 2, subunit 3 gamma
Gene Aliases:
Su(var)3-9, fi03g03, wu:fi03g03, wu:fi37d04, zgc:63805
Chromosome Location:
Chr.24: 16983426 - 17002950 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03139750_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eif2s3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212711.3 NP_997876.2 4 - 5 675 66
GenBank mRNA AY648723.1 - 4 - 5 675 66
BC055248.1 - 4 - 5 409 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
translation
translational initiation
positive regulation of translational fidelity
tRNA binding
nucleotide binding
translation initiation factor activity
GTPase activity
GTP binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0