This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03140065_m1
See other AGPAT3 GE Assays ›
Gene Symbol:
agpat3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
fd13g09, fj78c06, wu:fa16h03, wu:fd13g09, wu:fj78c06, wu:fq33b10, zgc:56524
Chromosome Location:
Chr.1: 46834730 - 46875494 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03140065_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: agpat3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213425.1 NP_998590.1 4 - 5 645 68
XM_005170976.4 XP_005171033.1 4 - 5 650 68
XM_005170977.4 XP_005171034.1 4 - 5 1020 68
XM_005170978.4 XP_005171035.1 4 - 5 601 68
XM_009293825.3 XP_009292100.1 5 - 6 852 68
XM_009293826.3 XP_009292101.1 4 - 5 1017 68
XM_009293837.3 XP_009292112.1 4 - 5 681 68
XM_009293841.3 XP_009292116.1 4 - 5 647 68
XM_021470311.1 XP_021325986.1 4 - 5 589 68
XM_021470376.1 XP_021326051.1 4 - 5 598 68
XM_021470393.1 XP_021326068.1 4 - 5 586 68
GenBank mRNA BC049474.1 - 4 - 5 645 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Process(es)

1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0