This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03143623_m1
See other ACMSD GE Assays ›
Gene Symbol:
acmsd
Gene Name:
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases:
zgc:162888
Chromosome Location:
Chr.9: 23244294 - 23253870 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03143623_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: acmsd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001089494.1 NP_001082963.1 3 - 4 289 57
GenBank mRNA BC139659.1 - 3 - 4 289 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

secondary metabolic process
negative regulation of quinolinate biosynthetic process
aminocarboxymuconate-semialdehyde decarboxylase activity
hydrolase activity
lyase activity
carboxy-lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0