This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03145133_m1
See other POLR3D GE Assays ›
Gene Symbol:
polr3d
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide D
Gene Aliases:
zgc:56407, zgc:77250
Chromosome Location:
Chr.Unknown: 4504 - 16419 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03145133_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: polr3d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200287.1 NP_956581.1 648 70
GenBank mRNA BC049426.1 - 648 70
BC066421.1 - 586 70
BC153604.1 - 508 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase III promoter
DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0