This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03146370_m1
See other STK25B GE Assays ›
Gene Symbol:
stk25b
Gene Name:
serine/threonine kinase 25b
Gene Aliases:
stk25, zgc:56022, zgc:77029
Chromosome Location:
Chr.2: 22617671 - 22636703 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03146370_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: stk25b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213308.1 NP_998473.1 3 - 4 619 82
XM_005171203.4 XP_005171260.1 3 - 4 662 82
GenBank mRNA BC045867.1 - 3 - 4 595 82
BC066512.1 - 3 - 4 619 82
BC164728.1 - 3 - 4 352 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
heart development
phosphorylation
blood vessel morphogenesis
protein kinase activity
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-qc9kr:80/100.66.79.73:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0