This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03147193_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
rbfox1
Gene Name:
RNA binding fox-1 homolog 1
Gene Aliases:
a2bp1, fox1, zgc:103635
Chromosome Location:
Chr.3: 27984158 - 28428193 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03147193_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rbfox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005596.1 NP_001005596.1 7 - 8 717 67
XM_017354041.2 XP_017209530.1 8 - 9 1065 67
XM_017354043.2 XP_017209532.1 7 - 8 787 67
XM_017354044.2 XP_017209533.1 8 - 9 1096 67
XM_017354045.2 XP_017209534.1 7 - 8 1323 67
XM_017354049.2 XP_017209538.1 7 - 8 949 67
XM_021470425.1 XP_021326100.1 8 - 9 944 67
XM_021470430.1 XP_021326105.1 8 - 9 1065 67
XM_021470431.1 XP_021326106.1 8 - 9 1065 67
XM_021470432.1 XP_021326107.1 8 - 9 1065 67
XM_021470435.1 XP_021326110.1 8 - 9 1065 67
XM_021470436.1 XP_021326111.1 8 - 9 1065 67
XM_021470437.1 XP_021326112.1 7 - 8 972 67
XM_021470439.1 XP_021326114.1 8 - 9 1065 67
XM_021470445.1 XP_021326120.1 8 - 9 1065 67
GenBank mRNA BC081500.1 - 7 - 8 717 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
hypothalamus development
regulation of RNA splicing
heart contraction
nucleic acid binding
RNA binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0