This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03149306_m1
See other MOGS GE Assays ›
Gene Symbol:
mogs
Gene Name:
mannosyl-oligosaccharide glucosidase
Gene Aliases:
gcs1, im:7160827, wu:fe50a12, wu:fk09a10, zgc:158312
Chromosome Location:
Chr.1: 41609226 - 41617118 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03149306_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mogs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080190.1 NP_001073659.1 3 - 4 815 110
XM_005160076.4 XP_005160133.1 4 - 5 902 110
GenBank mRNA BC129197.1 - 3 - 4 815 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
oligosaccharide metabolic process
catalytic activity
mannosyl-oligosaccharide glucosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline