This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03151499_m1
See other PITPNB GE Assays ›
Gene Symbol:
pitpnb
Gene Name:
phosphatidylinositol transfer protein, beta
Gene Aliases:
zgc:56092
Chromosome Location:
Chr.10: 4046376 - 4087018 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03151499_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pitpnb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001328435.1 NP_001315364.1 3 - 4 273 78
NM_200920.2 NP_957214.2 3 - 4 273 78
GenBank mRNA BC047829.1 - 3 - 4 273 78
BC165700.1 - 3 - 4 235 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

phospholipid transport
photoreceptor cell outer segment organization
phospholipid transporter activity
phosphatidylcholine transporter activity
phosphatidylinositol transporter activity
phosphatidylcholine binding
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline