This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03152081_m1
See other NTMT1 GE Assays ›
Gene Symbol:
ntmt1
Gene Name:
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases:
NTM1A, mettl11a, zgc:77488
Chromosome Location:
Chr.5: 1992 - 4681 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03152081_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ntmt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212898.1 NP_998063.1 3 - 4 472 69
GenBank mRNA BC067378.1 - 3 - 4 472 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid methylation
spindle organization
chromosome segregation
N-terminal peptidyl-glycine methylation
N-terminal peptidyl-proline dimethylation
methylation
N-terminal peptidyl-serine dimethylation
N-terminal peptidyl-serine trimethylation
methyltransferase activity
protein methyltransferase activity
transferase activity
histone methyltransferase activity
N-terminal protein N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline