This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03160069_s1
See other EIF5A GE Assays ›
Gene Symbol:
eif5a
Gene Name:
eukaryotic translation initiation factor 5A
Gene Aliases:
wu:fb17e01, wu:fb63h04, wu:fk81e04, zgc:77099
Chromosome Location:
Chr.24: 26481809 - 26485340 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03160069_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eif5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213185.1 NP_998350.1 4 1151 115
XM_005162775.3 XP_005162832.1 5 1220 115
XM_005162776.3 XP_005162833.1 5 1406 115
XM_005162777.2 XP_005162834.1 5 1169 115
GenBank mRNA BC056297.1 - 4 1133 115
BC067190.1 - 4 1151 115
BC152188.1 - 4 1138 115
BC152189.1 - 5 1181 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
positive regulation of translational elongation
positive regulation of translational termination
RNA binding
translation initiation factor activity
translation elongation factor activity
ribosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0