This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03165095_g1
See other SLC25A17 GE Assays ›
Gene Symbol:
slc25a17
Gene Name:
solute carrier family 25 member 17
Gene Aliases:
si:dkey-68o6.9, zgc:162641
Chromosome Location:
Chr.3: 158989 - 169920 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03165095_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc25a17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001099261.2 NP_001092731.2 9 3331 108
GenBank mRNA BC139873.1 - 3653 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

transmembrane transport
ATP transmembrane transporter activity
ADP transmembrane transporter activity
coenzyme A transmembrane transporter activity
FAD transmembrane transporter activity
FMN transmembrane transporter activity
NAD transporter activity
AMP transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0