This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03183833_s1
See other CHRAC1 GE Assays ›
Gene Symbol:
chrac1
Gene Name:
chromatin accessibility complex subunit 1
Gene Aliases:
wu:fb37b01, zgc:110753
Chromosome Location:
Chr.19: 41515776 - 41518918 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03183833_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: chrac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013293.1 NP_001013311.1 3 449 74
GenBank mRNA BC090283.1 - 3 449 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

primitive erythrocyte differentiation
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-m9fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline