This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03202042_s1
See other RAC1A GE Assays ›
Gene Symbol:
rac1a
Gene Name:
Rac family small GTPase 1a
Gene Aliases:
rac1, wu:fd16e02, zgc:55823, zgc:55917, zgc:86934
Chromosome Location:
Chr.12: 10464897 - 10476337 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03202042_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rac1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199771.1 NP_956065.1 6 1652 146
GenBank mRNA BC044501.1 - 6 1652 146
BC044538.1 - 6 1652 146
BC071548.1 - 6 1660 146
BC150471.1 - 3 1223 146

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
establishment or maintenance of cell polarity
small GTPase mediated signal transduction
myoblast fusion
motor neuron axon guidance
regulation of cell shape
Rac protein signal transduction
cell projection assembly
actin cytoskeleton organization
neutrophil chemotaxis
cortical cytoskeleton organization
regulation of actin cytoskeleton organization
cell migration involved in gastrulation
engulfment of apoptotic cell
regulation of neutrophil migration
nucleotide binding
GTPase activity
GTP binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9gdk9:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline