This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03421571_m1
See other RPL15 GE Assays ›
Gene Symbol:
rpl15
Gene Name:
ribosomal protein L15
Gene Aliases:
wu:fa92b05, zgc:92114
Chromosome Location:
Chr.19: 17385602 - 17393198 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03421571_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rpl15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003447.1 NP_001003447.1 1 - 2 186 72
XM_021468049.1 XP_021323724.1 1 - 2 210 72
GenBank mRNA BC075894.1 - 1 - 2 186 72
BC155095.1 - 1023 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translation
translation
RNA binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline