This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03421644_m1
See other SH3BGR GE Assays ›
Gene Symbol:
sh3bgr
Gene Name:
SH3 domain binding glutamate-rich protein
Gene Aliases:
zgc:100896
Chromosome Location:
Chr.15: 5923828 - 5968672 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03421644_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sh3bgr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003524.2 NP_001003524.1 1 - 2 185 70
NM_001362476.1 NP_001349405.1 1 - 2 185 70
NM_001362477.1 NP_001349406.1 1 - 2 185 70
NM_001362478.1 NP_001349407.1 1 - 2 185 70
NM_001362479.1 NP_001349408.1 1 - 2 185 70
NM_001362480.1 NP_001349409.1 1 - 2 185 70
XM_005169682.4 XP_005169739.1 1 - 2 210 70
XM_005169685.4 XP_005169742.1 1 - 2 210 70
XM_005169686.4 XP_005169743.1 1 - 2 173 70
XM_005169688.4 XP_005169745.1 1 - 2 170 70
XM_009291451.3 XP_009289726.1 1 - 2 210 70
XM_009291452.3 XP_009289727.1 1 - 2 210 70
XM_009291453.3 XP_009289728.1 1 - 2 210 70
XM_009291457.3 XP_009289732.1 1 - 2 180 70
XM_021481381.1 XP_021337056.1 1 - 2 210 70
XM_021481383.1 XP_021337058.1 1 - 2 180 70
XM_021481384.1 XP_021337059.1 1 - 2 180 70
XM_021481385.1 XP_021337060.1 1 - 2 169 70
GenBank mRNA BC077137.1 - 1 - 2 158 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline