This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03422367_m1
See other ERCC8 GE Assays ›
Gene Symbol:
ercc8
Gene Name:
excision repair cross-complementation group 8
Gene Aliases:
zgc:103550
Chromosome Location:
Chr.8: 90728 - 109706 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03422367_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ercc8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005984.1 NP_001005984.1 4 - 5 613 67
GenBank mRNA BC083423.1 - 4 - 5 613 67
BC154184.1 - 4 - 5 661 67
BC154372.1 - 4 - 5 665 67
BC165832.1 - 4 - 5 439 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
transcription-coupled nucleotide-excision repair
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA repair
ubiquitin-protein transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0